Learn More
Abnova™ AFP (Human) ELISA Kit
Sandwich enzyme immunoassay for the quantitative measurement of human AFP.
Manufacturer:  Abnova™ KA4792
Catalog No. 89-932-801
Add to cart
Description
Description
  • Used for the in vitro quantitative determination of human AFP in human serum, human plasma, cell lysate and buffered solution
  • Assay will recognize native and recombinant human AFP
Specifications
Specifications
ELISA Kit
P02771
AFP (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human AFP.
174
For research use only (RUO)
Store the kit at 4°C.
Buffered Solution, Cell Lysates, Plasma, Serum
Biotin
2 to 128 ng/mL
Quantification
Human Alpha Fetoprotein microtiter plate, Incubation Buffer, Washing Buffer (20x), Standard Protein, Standard/Sample Dilution Buffer, Secondary Antibody, AV-HRP, Secondary Antibody/AV-HRP Dilution Buffer, Substrate (Stabilized chromogen), Stop Solution: 1 N solution of sulfuric acid (H2SO4), Plate sealers: Adhesive sheet
100 μL
Human
Colorimetric