Learn More
Tocris Bioscience™ AG 045572
$111.30 - $236.50
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
27-301-0 View Documents Tocris Bioscience™
2730/10
10mg Each for $236.50
Add to cart
27-301-R View Documents Tocris Bioscience™
2730/1
Each for $111.30
Add to cart
Description
Description
Selective gonadotropin-releasing hormone (GnRH) receptor antagonist (Ki values are 2.2, 3.8 and 6.0 nM for mouse, rat and human receptors respectively). Suppresses testosterone and luteinising hormone (LH) levels in vivo.
Safety and Handling
Safety and Handling
Recommended Storage : Store at RT