Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AG 1478 hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent and selective inhibitor of epidermal growth factor receptor kinase (IC50 values 3 nM for EGFR and > 100 μM for ErbB2 and PDGFR). Inhibits proliferation of NCI-H2170 NSCLC cells in vitro (IC50 = 1 μM).
Specifications
Quantity | 10 mg |
Formula Weight | 352.22 |
Percent Purity | >98% |
Chemical Name or Material | AG 1478 hydrochloride |
Safety and Handling
Recommended Storage | Desiccate at -20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction