Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AG-2/AGR2 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP105936AF350
Description
AG-2/AGR2 Polyclonal specifically detects AG-2/AGR2 in Human, Mouse, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
AG-2/AGR2 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
AG-2, AG2member 17, anterior gradient 2 homolog (Xenopus laevis), anterior gradient homolog 2 (Xenopus laevis), GOB-4, hAG-2, HPC8, secreted cement gland homolog | |
Rabbit | |
0.1 mL | |
10551 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
AGR2 | |
Synthetic peptide made to an N-terminal portion of mouse AGR2 (within residues 1-50). [Swiss-ProtNo. O88312] | |
Primary | |
Human, Mouse, Bovine | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction