Learn More
Medchemexpress LLC Ag-494 | 133550-35-3 | MFCD00209863 | >98.0% | 280.28 g/mol | C16H12N2O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1010425MG
Encompass_Preferred
AG-494 is a small-molecule inhibitor of epidermal growth factor receptor (EGFR) tyrosine kinase used in biochemical and cellular research to probe EGFR-mediated signaling pathways. The compound is supplied as a solid for laboratory use and is intended for research purposes only.
  • Potent and selective EGFR tyrosine kinase inhibitor.
  • Blocks EGFR autophosphorylation in biochemical and cell-based assays.
  • Suitable for studying receptor-mediated signaling and inhibitor profiling.
  • Available in small-scale quantities suitable for laboratory experiments.
  • Provided as a solid with documented purity for research applications.
Catalog No. 50-002-12028