Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AG 99, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Epidermal growth factor receptor (EGFR) kinase inhibitor (IC50 = 10 μM) that is selective over insulin receptor kinase. Blocks tyrosine phosphorylation of p145met and promotes cell death of normal and cancer cells via activation of caspase-like proteases in vitro.
Specifications
Quantity | 50 mg |
Synonym | tyrphostin 46, tyrphostin a46, tyrphostin b40, alpha-cyano-3,4-dihydroxy cinnamide, 3,4-dihydroxy-alpha-cyanocinnamamide, tyrphostin ag 99, e-2-cyano-3-3,4-dihydroxyphenyl prop-2-enamide, 2-propenamide, 2-cyano-3-3,4-dihydroxyphenyl, 2e-2-cyano-3-3,4-dihydroxyphenyl prop-2-enamide, tyrphostin ag99 |
Formula Weight | 204.18 |
Percent Purity | >99% |
Chemical Name or Material | AG 99 |
Safety and Handling
Recommended Storage | Desiccate at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction