Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGAP11 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549230

Catalog No. PA549230


Add to cart

Description

Description

Putative GTPase-activating protein.
Specifications

Specifications

AGAP11
Polyclonal
PBS with 0.09% sodium azide
Q8TF27
Arf-GAP with GTPase, ANK repeat and PH domain-containing protein 11, AGAP-11, Centaurin-gamma-like protein KIAA1975, AGAP11, KIAA1975
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
119385
Western Blot
Unconjugated
AGAP11
Liquid
AGAP11
KLH conjugated synthetic peptide between 85-111 amino acids from the N-terminal region of human AGAP11.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit