Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGAP3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP286961

Catalog No. NB034583


Add to cart

Description

Description

AGAP3 Polyclonal specifically detects AGAP3 in Human samples. It is validated for Western Blot.
Specifications

Specifications

AGAP3
Unconjugated
PBS, 2% Sucrose
ANK repeat and PH domain-containing protein 3, ArfGAP with GTPase domain, ankyrin repeat and PH domain 3, centaurin-gamma-3, Cnt-g3, CRAG, CRAM-associated GTPase, CRMP (collapsin response mediator protein) associated, FLJ16146, FLJ34452, gamma 3, MR1-interacting protein, MRIP-1
The immunogen is a synthetic peptide directed towards the C-terminal region of Human AGAP3
100 μg
Signal Transduction
Primary
116988
Western Blot
Western Blot 1.0 ug/ml
Purified
Rabbit
Immunogen affinity purified
RUO
Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles.
Polyclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit