Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGAP8 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549343

Catalog No. PA549343

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Putative GTPase-activating protein.
Specifications

Specifications

AGAP8
Polyclonal
PBS with 0.09% sodium azide
Q96P64
Arf-GAP with GTPase, ANK repeat and PH domain-containing protein 8, AGAP-8, Centaurin-gamma-like family member 5, AGAP8, CTGLF5
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
119016
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
AGAP4
Liquid
AGAP4
KLH conjugated synthetic peptide between 498-531 amino acids from the C-terminal region of human AGAP8.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit