Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Agar

Used extensively as a gelling agent in preparation of culture media.

Manufacturer:  Thermo Scientific™ J10654Q1

 View more versions of this product

Catalog No. AAJ10654Q1


Order in Bulk

Request bulk or custom formats

Add to cart

Description

Description

  • Light tan granular powder
  • CAS: 9002-18-0
  • MDL Number: MFCD00081288
Specifications

Specifications

Light tan
Plastic Bottle
Agar
Granular Powder
25 lb.
Used as a gelling agent in preparation of culture media.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit