Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agar, Thermo Scientific™

Used extensively as a gelling agent in preparation of culture media.

$101.70 - $1422.00

Specifications

Product Type Agar
Format Granular Powder
For Use With (Application) Used as a Gelling Agent in Preparation of Culture Media.
Color Light tan
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ10654P2
View Documents
LAR NJ Thermo Scientific
J10654P2
1 lb.
Each for $101.70
Add to cart
 
AAJ10654P5
View Documents Promotion Details
LAR NJ Thermo Scientific
J10654P5
5 lb.
Each for $320.00
Add to cart
 
AAJ10654Q1
View Documents
LAR NJ Thermo Scientific
J10654Q1
25 lb.
Each for $1,422.00
Add to cart
 
Description

Description

  • Light tan granular powder
  • CAS: 9002-18-0
  • MDL Number: MFCD00081288
Specifications

Specifications

Agar
Granular Powder
Used as a Gelling Agent in Preparation of Culture Media.
Light tan
Safety and Handling

Safety and Handling

EINECSNumber : 232-658-1

Recommended Storage : Ambient temperatures

SDS
Product Certifications

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit