Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Agarose LE, Molecular Biology Grade, Ultrapure, Thermo Scientific™

Click to view available options
Quantity:
25g
100g
250g
500g
1kg
Packaging:
Plastic bottle
9012-36-6
MFCD00081294
9012-36-6
MFCD00081294
Packaging Plastic bottle
Physical Form Granular, free to flowing powder
Grade Molecular Biology
Quantity 1kg
Chemical Name or Material Agarose LE