Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Agarose, low EEO

Click to view available options
Quantity:
50g
100g
9012-36-6
MFCD00081294
9012-36-6
MFCD00081294
Applications
Agarose is useful in separation of nucleic acids electrophoretically, Suitable for isoelectric focusing, protein blotting and antibody separation Agarose gels are also used in immunoelectrophoresis (IEP) and double-diffusion Ouchterlony plates to demonstrate antibody-antigen reactions.

Solubility
Soluble in water

Notes
Gel strength: >2400g/cm² (1.5% w/v); Gelling temperature: 35-37°C; EEO: 0.07-0.1
Solubility Information Soluble in water
Physical Form Powder
Melting Point 87°C to 89°C
Color White
Quantity 100g
Sensitivity Moisture Sensitive & Hygroscopic
Chemical Name or Material Agarose, low EEO