Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agarose, PFGE, Ultrapure Bioreagent, J.T.Baker™

Click to view available options
Quantity:
100g
25g
Packaging:
Poly Bottle
POLYSTORMOR

Chemical Identifiers

9012-36-6

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Rapid Resolution of DNA Greater than 1,000 bp
CAS 9012-36-6
Packaging POLYSTORMOR
Quantity 25g
Specific Gravity 1L = 0.90kg
Chemical Name or Material Agarose, PFGE
Grade Ultrapure Bioreagent