Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agarose, Standard, Low Electroendosmosis (EEO), Ultrapure Bioreagent, J.T.Baker™

Click to view available options
Quantity:
100g
500g
Packaging:
Poly Bottle
9012-36-6
9012-36-6

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Electrophoresis
Packaging Poly Bottle
Grade Ultrapure Bioreagent
Quantity 100g
Specific Gravity 1L = 0.90kg
Chemical Name or Material Agarose, Standard, Low Electroendosmosis (EEO)