Learn More
Medchemexpress LLC Agarotetrol | 69809-22-9 | 100.0% | 318.32 g/mol | C17H18O6 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN1468R10MG
Encompass_Preferred
Agarotetrol (Standard) is a high-purity analytical reference standard of agarotetrol, a chromone derivative isolated from agarwood. Supplied as a solid for research and analytical use, it is intended for calibration, method development, and identity/purity confirmation in chromatographic and spectrometric analyses. Not for human or clinical use.
  • Analytical standard for calibration and method development.
  • Reported purity approximately 99.98%.
  • Molecular formula C17H18O6 and molecular weight 318.32 g/mol.
  • Available in small pack sizes suitable for analytical workflows.
  • Certificate of analysis and safety data sheet available for download.
Catalog No. 50-002-96560