Learn More
Abnova™ Ager (Mouse) ELISA Kit
Sandwich enzyme-linked immunosorbent assay
Manufacturer:  Abnova™ KA5728
Catalog No. 89-339-704
Add to cart
Description
Description
Ager (Mouse) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for the quantitative measurement of mouse Ager.
Specifications
Specifications
Quantitative
Ager
1 Kit
RUO
Mouse
11596
Standard curve
Store at 4°C for 6 months, at -20°C for 12 months. Avoid multiple freeze-thaw cycles.
RAGE