Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGER Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

$395.30

Specifications

Antigen AGER
Concentration 0.23 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-173-0885
SDP
Encompass_Preferred
View Documents
Proteintech Group Inc
163461AP150UL
150 μL
Each of 1 for $395.30
Add to cart
 
Description

Description

The receptor for advanced glycation endproducts (RAGE) is a member of the Immunoglobin superfamily of cell surface markers. This protein is able to interact with many molecules including: advanced glycation endproducts (AGE), amphoterin, and ligands. Research has shown that the accumulation of RAGE ligands in the biological system (endothelium mononuclear phagocytes, neurons and smooth muscle cells) can lead to destructive tissue diseases such as diabetic retinopathy, amyloidoses, tumors and inflammation disorders.
Specifications

Specifications

AGER
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
IgG
-20°C
0.23 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Antigen Affinity Chromatography
Antibody
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit