Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AGER Rat anti-Mouse, DyLight 755, Clone: RM0127-2B65, Novus Biologicals
Rat Monoclonal Antibody
Manufacturer: Novus Biologicals NBP122504IR
Description
AGER Monoclonal antibody specifically detects AGER in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
AGER | |
RM0127-2B65 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
Purified | |
AGER | |
Mouse recombinant protein of the extra cellular domain of RAGE protein | |
Protein G purified | |
Cancer | |
Primary | |
177 | |
Mouse |
Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
advanced glycosylation end product-specific receptor, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products | |
Rat | |
IgG2 | |
0.1 ml | |
Store at 4C in the dark. | |
Monoclonal | |
This was produced from a hybridoma (mouse myeloma) fused with spleen cells from a rat immunized with mouse recombinant protein of the extra cellular domain of RAGE protein. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title