Learn More
Abnova™ AGGF1 293T Cell Transient Overexpression Lysate (Denatured) (T02)
Human overexpression lysate
Manufacturer:  Abnova™ H00055109T02
Catalog No. 89-932-820
Add to cart
Description
Description
Specifications
Specifications
293T
BC032844.1
pCMV-AGGF1 full-length
100 μL
Human
AGGF1 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 78.65kDa
Transient overexpression cell lysate was tested with anti-AGGF1 antibody (Mfr. No. H00055109-B02) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot