Learn More
Abnova™ AGGF1 293T Cell Transient Overexpression Lysate (Denatured) (T04)
Human overexpression lysate
Manufacturer:  Abnova™ H00055109T04
Catalog No. 89-932-819
Add to cart
Description
Description
Specifications
Specifications
293T
BC002828.2
pCMV-AGGF1 full-length
100 μL
Human
AGGF1 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 12.1kDa
Transient overexpression cell lysate was tested with anti-AGGF1 antibody (Mfr. No. H00055109-B03) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot