Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aggrecan Rabbit anti-Human,Mouse, Unconjugated, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$184.00 - $463.00
Specifications
Antigen | Aggrecan |
---|---|
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABT54525UMI
|
MilliporeSigma
ABT54525UL |
25 μL |
Each for $184.00
|
|
|||||
ABT545100MI
|
MilliporeSigma
ABT545100UL |
100 μL |
Each for $463.00
|
|
|||||
Description
Aggrecan Polyclonal antibody specifically detects Aggrecan in Human,Mouse samples. It is validated for Western Blotting, Immunohistochemistry (Paraffin).Specifications
Aggrecan | |
Polyclonal | |
Purified | |
Cell Structure | |
NP_031450 | |
Acan, Agc, Agc1 | |
IgG | |
Affinity Purified | |
Anti-Aggrecan is a rabbit polyclonal antibody that detects Aggrecan core protein. It targets an epitope within 150 amino acids from the C-terminal half. |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Aggrecan core protein;Cartilage-specific proteoglycan core protein;CSPCP | |
GST/His-tagged recombinant fragment corresponding to 150 amino acids from the C-terminal half of murine Aggrecan core protein. | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
∼270 kDa observed; 221.94 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title