Learn More
Tocris Bioscience™ Agitoxin 2
Potent Shaker K+ channel blocker
Manufacturer:  Tocris Bioscience™ 5195/10U
Catalog No. 51-951-0U
Due to product restrictions, please to purchase or view availability for this product.
This item is not returnable. View return policy
Description
Description
Potent Shaker K+ channel blocker (Ki = 0.64 nM). Also inhibits Kv1.3, Kv1.6 and Kv1.1 K+ channels (Ki values are 4, 37 and 44 pM respectively).
Specifications
Specifications
Agitoxin 2
Agitoxin 2
4090.87
10μg