Learn More
AGN 192403 hydrochloride, Tocris
I1 selective ligand
Manufacturer:  R&D Systems 1072/10
Quantity 10mg
 View more versions of this product
Catalog No. 10-721-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>98%
189.73
AGN 192403 hydrochloride
C10H19N·HCl
(±)-2-endo-Amino-3-exo-isopropylbicyclo[2.2.1]heptane hydrochloride
Specifications
1021868-90-5
C10H19N·HCl
10mg
AGN 192403 hydrochloride
>98%
189.73
(±)-2-endo-Amino-3-exo-isopropylbicyclo[2.2.1]heptane hydrochloride
Safety and Handling
Recommended Storage : Store at Room Temperature