Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ago1/2/3 Monoclonal Antibody (2A8), Active Motif™
Mouse Monoclonal Antibody
$147.50 - $576.30
Specifications
Antigen | Ago1/2/3 |
---|---|
Clone | 2A8 |
Concentration | 1 mg/mL |
Applications | Immunoprecipitation |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-2980
|
Active Motif
39938 |
10 μg |
Each for $147.50
|
|
|||||
50-199-2979
|
Active Motif
39937 |
100 μg |
Each for $576.30
|
|
|||||
Description
The Argonaute family of proteins play roles in RNA interference and in short-interfering-RNA-mediated gene silencing. Small interfering RNAs (siRNAs) have gained much attention for their powerful ability to suppress gene expression. Introduction of double-stranded RNA (dsRNA), that are homologues in sequence to a gene, has proven to suppress that gene's expression through a process known as RNA interference (RNAi); this process post-transcriptionally silences a gene through mRNA inhibition or degradation. Both biochemical and genetic studies have led to the identification of two, conserved gene families that are universal components of the interference process. These are the Dicer family, which is comprised of members of the Ribonuclease III family of enzymes, and the Argonaute gene family. Argonaute proteins addition to their role in RNAi process, may also influence development, such as, stem cell fate determination.Specifications
Ago1/2/3 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8 | |
192669, 26523, 27161 | |
This Ago1/2/3 antibody was raised against a recombinant protein corresponding to amino acids 47-859 of human Ago2. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
AGO2 |
2A8 | |
Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human | |
Q9H9G7, Q9UKV8, Q9UL18 | |
AGO1, AGO2, AGO3 | |
IgG | |
Protein G | |
Antibody |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title