Learn More
Ago2/eIF2C2 Mouse anti-Human, Mouse, Clone: 2A8, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259159
Catalog No. NBP259159
Add to cart
Description
Description
Ago2/eIF2C2 Monoclonal antibody specifically detects Ago2/eIF2C2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunofluorescence.
Specifications
Specifications
Ago2/eIF2C2
2A8
Western Blot 1:300-1:500, Immunohistochemistry 1:500, Immunoprecipitation, Immunofluorescence 1:500
Purified
EIF2C2
The exact immunogen is propietary information.
Protein A purified
RUO
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunofluorescence
Unconjugated
PBS with 0.05% Sodium Azide
AGO2argonaute 2, argonaute2, EC 3.1.26.n1, EC 3.1.26.n2, eIF2C 2, eIF-2C 2, Eukaryotic translation initiation factor 2C 2, eukaryotic translation initiation factor 2C, 2, hAgo2, MGC3183, PAZ Piwi domain protein, PPD, protein argonaute-2, Protein slicer, Q10
Mouse
IgG1
100 ug
Epigenetics, microRNA
Primary
27161