Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ago2/eIF2C2 Rabbit anti-Human, Janelia Fluor 646, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP299647JF646

Catalog No. NB062283


Add to cart

Description

Description

Ago2/eIF2C2 Polyclonal antibody specifically detects Ago2/eIF2C2 in Human samples. It is validated for Immunofluorescence
Specifications

Specifications

Ago2/eIF2C2
Janelia Fluor 646
50mM Sodium Borate
AGO2argonaute 2, argonaute2, EC 3.1.26.n1, EC 3.1.26.n2, eIF2C 2, eIF-2C 2, Eukaryotic translation initiation factor 2C 2, eukaryotic translation initiation factor 2C, 2, hAgo2, MGC3183, PAZ Piwi domain protein, PPD, protein argonaute-2, Protein slicer, Q10
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human Ago2/eIF2C2.
Antigen Affinity-purified
RUO
Store at 4°C in the dark.
Polyclonal
Human
Immunofluorescence
Immunocytochemistry/Immunofluorescence
Purified
Rabbit
IgG
0.1 mL
Epigenetics
Primary
27161
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit