Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGPAT4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549627

Catalog No. PA549627

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with monkey based on sequence homology.

This gene encodes a member of the 1-acylglycerol-3-phosphate O-acyltransferase family. This integral membrane protein converts lysophosphatidic acid to phosphatidic acid, the second step in de novo phospholipid biosynthesis. [provided by RefSeq, Jul 2008].
Specifications

Specifications

AGPAT4
Polyclonal
Unconjugated
AGPAT4
Liquid
AGPAT4
KLH conjugated synthetic peptide between 176-202 amino acids from the Central region of human AGPAT4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q9NRZ5
1-acyl-sn-glycerol-3-phosphate acyltransferase delta, 1-acylglycerol-3-phosphate O-acyltransferase 4, 1-AGP acyltransferase 4, 1-AGPAT 4, Lysophosphatidic acid acyltransferase delta, LPAAT-delta, AGPAT4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
56895
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit