Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGPAT5 Mouse anti-Human, Clone: OTI5F4, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA504143

Catalog No. 50-167-4892


Add to cart

Description

Description

This gene encodes a member of the 1-acylglycerol-3-phosphate O-acyltransferase family. This integral membrane protein converts lysophosphatidic acid to phosphatidic acid, the second step in de novo phospholipid biosynthesis.
Specifications

Specifications

AGPAT5
Monoclonal
0.88 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9NUQ2
1AGPAT5, LPAATE
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55326
Western Blot
OTI5F4
Unconjugated
AGPAT5
Liquid
AGPAT5
Full length human recombit protein of human AGPAT5 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit