Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AGPAT5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549935
Description
The antibody detects endogenous levels of total AGPAT5 protein.
AGPAT5 Polyclonal antibody specifically detects AGPAT5 in Human samples. It is validated for Western Blot.Specifications
AGPAT5 | |
Polyclonal | |
Unconjugated | |
AGPAT5 | |
1110013A05Rik; 1-acylglycerol-3-phosphate O-acyltransferase 5; 1-acylglycerol-3-phosphate O-acyltransferase 5 (lysophosphatidic acid acyltransferase, epsilon); 1-acyl-sn-glycerol-3-phosphate acyltransferase epsilon; 1-AGP acyltransferase 5; 1-AGPAT 5; 1AGPAT5; 1-AGPAT5; AGPAT5; D8Ertd319e; EC 2.3.1.51; LPAATE; LPAAT-epsilon; lysophosphatidic acid acyltransferase epsilon; RGD1306405; testicular tissue protein Li 144 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55326 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9NUQ2 | |
AGPAT5 | |
Synthesized peptide derived from internal of human AGPAT5. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction