Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGPAT5 Mouse anti-Human, Clone: OTI1H10, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525829

Catalog No. PIMA525829


Add to cart

Description

Description

This gene encodes a member of the 1-acylglycerol-3-phosphate O-acyltransferase family. This integral membrane protein converts lysophosphatidic acid to phosphatidic acid, the second step in de novo phospholipid biosynthesis.
Specifications

Specifications

AGPAT5
Monoclonal
0.66 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9NUQ2
1AGPAT5, LPAATE, 1-AGP acyltransferase 5, 1-AGPAT 5, 1-acyl-sn-glycerol-3-phosphate acyltransferase epsilon, lysophosphatidic acid acyltransferase epsilon
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55326
Immunohistochemistry (Paraffin), Western Blot
OTI1H10
Unconjugated
AGPAT5
Liquid
AGPAT5
Full length human recombit protein of AGPAT5 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit