Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGPS Mouse anti-Human, FITC, Clone: AGPS-03, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP262219F

Catalog No. NB002710


Add to cart

Description

Description

AGPS Monoclonal specifically detects AGPS in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

AGPS
AGPS-03
Flow Cytometry
Purified, Conjugated
AGPS
recombinant human AGPS (amino acids 158-384)
Protein A purified
Store at 4°C in the dark.
Monoclonal
The antibody MEM-154 reacts with an epitope on CD16 antigen that is residing in proximity to FG loop (probably BC or C'E loop). CD16 is a low affinity receptor for aggregated IgG (FcgammaRIII antigen). The antibody MEM-154 reacts with CD16+ granulocytes.
Flow Cytometry
FITC
PBS with 0.05% Sodium Azide
ADAP-S, ADAS, ADHAPSAlkylglycerone-phosphate synthase, ADPS, Aging-associated gene 5 protein, aging-associated protein 5, ALDHPSY, Alkyl-DHAP synthase, alkyldihydroxyacetonephosphate synthase, peroxisomal, alkylglycerone phosphate synthase, DKFZp762O2215, EC 2.5.1.26, FLJ99755
Mouse
IgG2a
0.1 mL
Primary
8540
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit