Learn More
AGTPBP1 Mouse anti-Human, Clone: OTI9C8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF800287

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
NNA1 is a zinc carboxypeptidase that contains nuclear localization signals and an ATP/GTP-binding motif that was initially cloned from regenerating spinal cord neurons of the mouse.Specifications
AGTPBP1 | |
Monoclonal | |
Unconjugated | |
AGTPBP1 | |
CCP1, NNA1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
23287 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI9C8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9UPW5 | |
AGTPBP1 | |
Human recombit protein fragment corresponding to amino acids 368-753 of human AGTPBP1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.