Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGTR-1 Mouse anti-Human, Clone: 1010103, R&D Systems™

Mouse Monoclonal Antibody

$128.24 - $329.00

Specifications

Antigen AGTR-1
Clone 1010103
Applications Flow Cytometry, CyTOF
Host Species Mouse
Target Species Human
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB10244SP
View Documents Promotion Details
R&D Systems
MAB10244SP
25 μg
Each for $128.24
Add to cart
 
MAB10244100
View Documents Promotion Details
R&D Systems
MAB10244100
100 μg
Each for $329.00
Add to cart
 
Description

Description

AGTR-1 Monoclonal specifically detects AGTR-1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications

Specifications

AGTR-1
Flow Cytometry, CyTOF
Human
Primary
1010103
Mouse
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit