Learn More
AGTR-1 Rabbit anti-AGTR-1, DyLight 550, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP177078R
Catalog No. NBP177078R
Add to cart
Description
Description
AGTR-1 Polyclonal antibody specifically detects AGTR-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
AGTR-1
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
Affinity Purified
AGTR1
Antibody was raised against a 16 amino acid synthetic peptide from near the center of human AGTR1. The immunogen is located within amino acids 210 - 260 of AGTR1.
Peptide affinity purified
Store at 4C in the dark.
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
D3DNG8
AG2S, AGTR1AAngiotensin II type-1 receptor, Angiostensin Receptor, angiotensin II receptor, type 1, angiotensin receptor 1, angiotensin receptor 1B, AT1AT1AR, AT1B, AT1R, AT2R1A, AT2R1AGTR1B, AT2R1BAT1BR, HAT1R, type-1 angiotensin II receptor, type-1B angiotensin II receptor
Rabbit
IgG
0.1 ml
Primary
185