Learn More
AGTR-2 Rabbit anti-Human, Mouse, Rat, DyLight 405, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP177368V
Catalog No. NBP177368V
Add to cart
Description
Description
AGTR-2 Polyclonal antibody specifically detects AGTR-2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
AGTR-2
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
Angiostensin Receptor, angiotensin II receptor, type 2, Angiotensin II type-2 receptor, angiotensin receptor 2, AT2ATGR2, MRX88type-2 angiotensin II receptor
Rabbit
IgG
0.1 ml
Apoptosis
Primary
186
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Affinity Purified
AGTR2
Antibody was raised against a 16 amino acid synthetic peptide from near the center of human AGTR2. The immunogen is located within amino acids 220 - 270 of AGTR2.
Peptide affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Human, Mouse, Rat