Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGTR-2 Rabbit anti-Human, Mouse, Rat, PE, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP177368PE

Catalog No. NBP177368PE


Add to cart

Description

Description

AGTR-2 Polyclonal antibody specifically detects AGTR-2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

AGTR-2
PE
PBS with 0.05% Sodium Azide
Angiostensin Receptor, angiotensin II receptor, type 2, Angiotensin II type-2 receptor, angiotensin receptor 2, AT2ATGR2, MRX88type-2 angiotensin II receptor
Rabbit
IgG
0.1 ml
Apoptosis
Primary
186
Western Blot
Western Blot
Affinity Purified
AGTR2
Antibody was raised against a 16 amino acid synthetic peptide from near the center of human AGTR2. The immunogen is located within amino acids 220 - 270 of AGTR2.
Peptide affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only