Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGTRAP Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549010

Catalog No. PA549010

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Appears to be a negative regulator of type-1 angiotensin II receptor-mediated signaling by regulating receptor internalisation as well as mechanism of receptor desensitization such as phosphorylation. Induces also a decrease in cell proliferation and angiotensin II-stimulated transcriptional activity.
Specifications

Specifications

AGTRAP
Polyclonal
PBS with 0.09% sodium azide
Q6RW13
Type-1 angiotensin II receptor-associated protein, AT1 receptor-associated protein, AGTRAP, ATRAP
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
57085
Western Blot
Unconjugated
AGTRAP
Liquid
AGTRAP
KLH conjugated synthetic peptide between 133-159 amino acids from the C-terminal region of human AGTRAP.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit