Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGXT2L1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP15496520UL

 View more versions of this product

Catalog No. NBP15496520


Add to cart

Description

Description

AGXT2L1 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

AGXT2L1
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
AGXT2L1
Synthetic peptides corresponding to AGXT2L1(alanine-glyoxylate aminotransferase 2-like 1) The peptide sequence was selected from the middle region of AGXT2L1. Peptide sequence KRVLLSADGPHRNVLKIKPPMCFTEEDAKFMVDQLDRILTVLEEAMGTKT.
56 kDa
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Western Blot
Western Blot 1:100-1:2000
Q8TBG4
alanine--glyoxylate aminotransferase 2-like 1, alanine-glyoxylate aminotransferase 2-like 1, EC 2.6.1, EC 2.6.1.-, EC 2.6.1.11, EC 2.6.1.44
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
64850
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit