Learn More
AH 11110 hydrochloride, Tocris
Subtype-selective α1B ligand
Manufacturer:  R&D Systems 0998/10
Quantity 10mg
 View more versions of this product
Catalog No. 09-981-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>98%
374.91
AH 11110 hydrochloride
C21H26N2O2·HCl
4-Imino-1-(2-phenylphenoxy)-4-piperidinebutan-2-ol hydrochloride
Specifications
179388-65-9
C21H26N2O2·HCl
4-Imino-1-(2-phenylphenoxy)-4-piperidinebutan-2-ol hydrochloride
AH 11110 hydrochloride
>98%
374.91
10mg
Safety and Handling
Recommended Storage : Store at Room Temperature