Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AHCTF1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549366

Catalog No. PA549366

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Required for the assembly of a functional nuclear pore complex (NPC) on the surface of chromosomes as nuclei form at the end of mitosis. May initiate NPC assembly by binding to chromatin and recruiting the Nup107-160 subcomplex of the NPC. Also required for the localization of the Nup107-160 subcomplex of the NPC to the kinetochore during mitosis and for the completion of cytokinesis.
Specifications

Specifications

AHCTF1
Polyclonal
PBS with 0.09% sodium azide
Q8WYP5
Protein ELYS, Embryonic large molecule derived from yolk sac, Protein MEL-28, Putative AT-hook-containing transcription factor 1, AHCTF1, ELYS, TMBS62
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
25909
Western Blot
Unconjugated
AHCTF1
Liquid
AHCTF1
KLH conjugated synthetic peptide between 1210-1260 amino acids from human.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit