Learn More
AHCY Mouse anti-Human, MaxPab™ Polyclonal , Abnova™
Mouse polyclonal antibody raised against a full-length human AHCY protein.
Supplier: Abnova Corporation H00000191B01
Description
S-adenosylhomocysteine hydrolase belongs to the adenosylhomocysteinase family. It catalyzes the reversible hydrolysis of S-adenosylhomocysteine (AdoHcy) to adenosine (Ado) and L-homocysteine (Hcy). Thus, it regulates the intracellular S-adenosylhomocysteine (SAH) concentration thought to be important for transmethylation reactions. Deficiency in this protein is one of the different causes of hypermethioninemia. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MSDKLPYKVADIGLAAWGRKALDIAENEMPGLMRMRERYSASKPLKGARIAGCLHMTVETAVLIETLVTLGAEVQWSSCNIFSTQDHAAAAIAKAGIPVYAWKGETDEEYLWCIEQTLYFKDGPLNMILDDGGDLTNLIHTKYPQLLPGIRGISEETTTGVHNLYKMMANGILKVPAINVNDSVTKSKFDNLYGCRESLIDGIKRATDVMIAGKVAVVAGYGDVGKGCAQALRGFGARVIITEIDPINALQAAMEGYEVTTMDEACQEGNIFVTTTGCIDIILGRHFEQMKDDAIVCNIGHFDVEIDVKWLNENAVEKVNIKPQVDRYRLKNGRRIILLAEGRLVNLGCAMGHPSFVMSNSFTNQVMAQIELWTHPDKYPVGVHFLPKKLDEAVAEAHLGKLNVKLTKLTEKQAQYLGMSCDGPFKPDHYRYSpecifications
AHCY | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human AHCY protein. | |
NM_000687.1 | |
AHCY | |
AHCY (NP_000678.1, 1 a.a. ∽ 432 a.a) full-length human protein. | |
RUO | |
191 | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
No additive | |
SAHH | |
Mouse | |
50 μL | |
Primary | |
Antibody reactive against mammalian transfected lysate. | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.