Learn More
AHCY Mouse anti-Human, Mouse, Clone: 445CT13.3.3, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA524721
Catalog No. PIMA524721
Add to cart
Description
Description
S-adenosylhomocysteine hydrolase (AHCY) catalyzes the reversible hydrolysis of S-adenosylhomocysteine (AdoHcy) to adenosine (Ado) and L-homocysteine (Hcy). Thus, it regulates the intracellular S-adenosylhomocysteine (SAH) concentration thought to be important for transmethylation reactions. Deficiency in this protein is one of the different causes of hypermethioninemia. S-adenosylhomocysteine hydrolase belongs to the adenosylhomocysteinase family.
Specifications
Specifications
AHCY
Monoclonal
Unconjugated
AHCY
Liquid
AHCY
KLH conjugated synthetic peptide between 79-110 amino acids from human AHCY
100 μL
-20°C
Primary
191, 269378
Western Blot
445CT13.3.3
ascites with 0.09% sodium azide
P23526, P50247
Adenosylhomocysteinase, AdoHcyase, S-adenosyl-L-homocysteine hydrolase, AHCY, SAHH
Mouse
IgM
RUO
Antibody
Monoclonal
Human, Mouse