Learn More
Abnova™ AHDC1 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00027245T01
Catalog No. 89-932-897
Add to cart
Description
Description
Specifications
Specifications
293T
BC014394
pCMV-AHDC1 full-length
100 μL
Human
AHDC1 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 41.7kDa
Transient overexpression cell lysate was tested with anti-AHDC1 antibody (Mfr. No. H00027245-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot