Learn More
Ahlstrom-Munksjo Qualitatitve Non Wet Strengthened Filter Paper, Grade 613
Used for the filtration of coarse particles and gelatinous precipitates such as ferric aluminum and magnesium hydroxides. Also used in air monitoring and gas filtration when high flow rate is needed.
Manufacturer:  Ahlstrom-Munksjo 61312020
Catalog No. 09-790-72
Add to cart
Description
Description
Specifications
Specifications
Sheet
613
Studeny use and general purpose lab filtration tasks
20 x 20 in.
Medium