Learn More
Ahlstrom-Munksjo Chromatography, Electrophoresis and Blotting Paper, Grade 222
Offers extremely high flow and is recommended for both electrophoresis of large molecules and as a blotter to complete the blotting sandwich in gel transfer applications.
Supplier:  Ahlstrom-Munksjo 22288105
 View more versions of this product
Catalog No. 09-790-48
Add to Cart
Description
Description
Specifications
Specifications
High
222
8 cm
Electrophoresis of large molecules and as blotter to complete the blotting sandwich in gel transfer
0.83 mm
10.5 cm
Product Suggestions
Product Suggestions