Learn More
Ahlstrom™ Filter Papers – Grade 222
Manufacturer:  Ahlstrom™ 22281015
 View more versions of this product
Catalog No. 09-790-49E
Add to cart
Specifications
Specifications
222
15cm
High
10cm
Electrophoresis of large molecules and as blotter to complete the blotting sandwich in gel transfer
0.83mm