Learn More
Ahlstrom™ Filter Papers – Grade 222
Manufacturer:  Ahlstrom™ 22284657
 View more versions of this product
Catalog No. 09-790-49K
Add to cart
Specifications
Specifications
222
57cm
High
46cm
Electrophoresis of large molecules and as blotter to complete the blotting sandwich in gel transfer
0.83mm