Learn More
Ahlstrom™ Filter Papers – Grade 238
Manufacturer:  Ahlstrom™ 23881015
 View more versions of this product
Catalog No. 22-279533
Add to cart
Specifications
Specifications
238
17cm
0.35mm
10cm
Chromatography applications. Use as wicks in protein or nucleic acid blotting. Suited for electrophoresis