Learn More
Ahlstrom™ Filter Papers – Grade 238 PROMO
Supplier:  Ahlstrom 23880710PM
 View more versions of this product
Catalog No. 0979053BPM
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
Specifications
Specifications
Chromatography applications. Use as wicks in protein or nucleic acid blotting. Suited for electrophoresis
0.35 mm
10 cm
238
7 cm
Product Suggestions
Product Suggestions